Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Galnt13
- Entrez Gene ID
-
271786
- Gene Name
- UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13
- Gene Aliases
-
A230002A12, A230020F20, BB182356
- Chromosome Location
-
Chr.2: 54436387 - 55118309 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC131652.1 |
- |
1 - 2 |
128 |
54 |
View Full Product Details