Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pramel6
- Entrez Gene ID
-
347711
- Gene Name
- preferentially expressed antigen in melanoma like 6
- Gene Aliases
-
C80453, C80515
- Chromosome Location
-
Chr.2: 87506565 - 87510868 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK139830.1 |
- |
1 - 2 |
212 |
94 |
View Full Product Details