Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E130119H09Rik
Entrez Gene ID
78550
Gene Name
RIKEN cDNA E130119H09 gene
Gene Aliases
-
Chromosome Location
Chr.13: 53912263 - 53916212 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK021396.1 - 1 - 2 256 84


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY731386.1 - 1 - 2 256 Chr. 13 - 53912644 - 53915748 [-] on Build GRCm38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream RIKEN cDNA 4930555G21 gene 4930555G21Rik -263.4kb Anti-sense Chr. - 53648585 - 53650080 on Build GRCm38
Downstream dopamine receptor D1 Drd1 C030036C15Rik, Drd-1, Drd1a, Gpcr15 135.4kb Sense Chr. - 54051183 - 54055658 on Build GRCm38

View Full Product Details