The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010067.4 | NP_034197.3 | 1 - 2 | 93 | 69 |
GenBank mRNA | AF012129.1 | - | 1 - 2 | 80 | 69 |
AF045889.1 | - | 1 - 2 | 85 | 69 | |
AK050403.1 | - | 1 - 2 | 93 | 69 | |
BC087892.1 | - | 1 - 2 | 93 | 69 |