The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082476.2 | NP_001075945.1 | 11 - 12 | 1502 | 56 |
NM_001252541.1 | NP_001239470.1 | 9 - 10 | 1301 | 56 | |
NM_001252542.1 | NP_001239471.1 | 11 - 12 | 1569 | 56 | |
GenBank mRNA | AK019617.1 | - | 11 - 12 | 1499 | 56 |
AK143124.1 | - | 11 - 12 | 1543 | 56 | |
AK163207.1 | - | 9 - 10 | 1271 | 56 |