The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026697.3 | NP_080973.1 | 7 - 8 | 842 | 82 |
GenBank mRNA | AB232609.1 | - | 6 - 7 | 473 | 82 |
AJ968955.1 | - | 7 - 8 | 697 | 82 | |
AJ968956.1 | - | 6 - 7 | 643 | 82 | |
AJ968957.1 | - | 7 - 8 | 697 | 82 | |
AJ968958.1 | - | 6 - 7 | 643 | 82 | |
AJ968959.1 | - | 7 - 8 | 697 | 82 | |
AJ968960.1 | - | 6 - 7 | 643 | 82 | |
AK002704.1 | - | 7 - 8 | 714 | 82 | |
AK133963.1 | - | 7 - 8 | 742 | 82 | |
AK135751.1 | - | 7 - 8 | 732 | 83 | |
AK160859.1 | - | 7 - 8 | 842 | 82 | |
AK165812.1 | - | 7 - 8 | 689 | 82 | |
AK168551.1 | - | 7 - 8 | 699 | 82 | |
AK168867.1 | - | 7 - 8 | 711 | 82 | |
AK169396.1 | - | 7 - 8 | 713 | 82 | |
AK216803.1 | - | 4 - 5 | 260 | 82 | |
BC009085.1 | - | 7 - 8 | 525 | 82 | |
BC025139.1 | - | 7 - 8 | 713 | 82 | |
BC056648.1 | - | 7 - 8 | 715 | 82 |