Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arl5b
Entrez Gene ID
75869
Gene Name
ADP-ribosylation factor-like 5B
Gene Aliases
4930587A11Rik, AW146430, Arl8
Chromosome Location
Chr.2: 15055362 - 15079191 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029466.4 NP_083742.2 1 - 2 217 67
GenBank mRNA AK016359.1 - 1 - 2 174 67
AK040422.1 - 1 - 2 260 67
AK041237.1 - 1 - 2 248 67
AK042200.1 - 1 - 2 303 67
AK053731.1 - 1 - 2 218 67
AK164341.1 - 1 - 2 212 67
BC066810.1 - 1 - 2 256 67

View Full Product Details