Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Smek1
Entrez Gene ID
68734
Gene Name
SMEK homolog 1, suppressor of mek1 (Dictyostelium)
Gene Aliases
1110034C04Rik, BC064465, mKIAA2010
Chromosome Location
Chr.12: 101039409 - 101083702 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160214.1 NP_001153686.1 5 - 6 1823 86
NM_211355.2 NP_997594.1 5 - 6 1823 86
GenBank mRNA AK051239.1 - 5 - 6 1748 86
AK152480.1 - 5 - 6 1820 86
AK156272.1 - 5 - 6 1785 86
AK173331.1 - 5 - 6 1726 86
BC064465.1 - 5 - 6 1521 86

View Full Product Details