Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galc
Entrez Gene ID
14420
Gene Name
galactosylceramidase
Gene Aliases
2310068B06Rik, A930008M05Rik, AW212969, AW413532, Gacy, twi, twitcher
Chromosome Location
Chr.12: 98202300 - 98259461 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK077475.1 - 7 - 8 721 67
AK154760.1 - 7 - 8 882 67
AK161339.1 - 2 - 3 172 67
BC065140.1 - 7 - 8 755 67
BC086671.1 - 7 - 8 759 67
D38557.1 - 7 - 8 829 67

View Full Product Details