The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029617.2 | NP_083893.2 | 18 - 19 | 5865 | 116 |
GenBank mRNA | AK017750.1 | - | 6 - 7 | 514 | 116 |