Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casc5
Entrez Gene ID
76464
Gene Name
cancer susceptibility candidate 5
Gene Aliases
2310043D08Rik, 5730505K17Rik
Chromosome Location
Chr.2: 119047038 - 119104829 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029617.2 NP_083893.2 3 - 4 190 157

View Full Product Details