Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfb1m
Entrez Gene ID
224481
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
AI429207
Chromosome Location
Chr.17: 3519256 - 3557822 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146074.1 NP_666186.1 3 - 4 395 147
GenBank mRNA AF508971.1 - 3 - 4 395 147
BC032930.1 - 3 - 4 395 147

View Full Product Details