Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmr1
Entrez Gene ID
14265
Gene Name
fragile X mental retardation syndrome 1
Gene Aliases
FMRP, Fmr-1
Chromosome Location
Chr.X: 68678541 - 68717963 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290424.1 NP_001277353.1 3 - 4 473 98
NM_008031.3 NP_032057.2 3 - 4 473 98
GenBank mRNA AF179463.1 - - 77 98
AF461114.1 - 3 - 4 197 98
AK046605.1 - 3 - 4 433 98
AK053701.1 - 3 - 4 433 98
AK053829.1 - 3 - 4 475 98
AK080948.1 - 3 - 4 433 98
AK208300.1 - 3 - 4 173 98
BC079671.1 - 3 - 4 459 98
L23971.1 - 3 - 4 320 98

View Full Product Details