Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankle1
Entrez Gene ID
234396
Gene Name
ankyrin repeat and LEM domain containing 1
Gene Aliases
8430438L13Rik, Ankrd41, C430003I23, Lem3
Chromosome Location
Chr.8: 71406005 - 71410543 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172756.2 NP_766344.2 8 - 9 1455 62
GenBank mRNA AK049395.1 - 6 - 7 1762 62
AK078830.1 - 7 - 8 1685 62
AK083552.1 - 6 - 7 1754 62
BC030436.1 - 8 - 9 1394 62
BC094316.1 - 8 - 9 1412 62
BC145659.1 - 4 - 5 678 62
EU184014.1 - 8 - 9 1431 62

View Full Product Details