The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029865.2 | NP_084141.2 | 1 - 2 | 695 | 61 |
GenBank mRNA | AK020528.1 | - | 1 - 2 | 409 | 61 |
AK155219.1 | - | 1 - 2 | 655 | 61 | |
BC019407.1 | - | 1 - 2 | 651 | 61 |