The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145802.1 | NP_001139274.1 | 1 - 2 | 103 | 70 |
NM_001145803.1 | NP_001139275.1 | 1 - 2 | 103 | 70 | |
GenBank mRNA | BC050009.1 | - | 1 - 2 | 42 | 70 |