Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dgkk
Entrez Gene ID
331374
Gene Name
diacylglycerol kinase kappa
Gene Aliases
Gm360
Chromosome Location
Chr.X: 6779306 - 6779636 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177914.3 NP_808582.3 - 2359 87
GenBank mRNA BC075627.1 - - 2359 87

View Full Product Details