The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025282.3 | NP_079558.1 | 10 - 11 | 1645 | 78 |
GenBank mRNA | AK009139.1 | - | 10 - 11 | 1485 | 78 |
AK077603.1 | - | 9 - 10 | 1446 | 78 | |
BC026841.1 | - | 10 - 11 | 1407 | 78 |