Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Btk
Entrez Gene ID
12229
Gene Name
Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
AI528679, xid
Chromosome Location
Chr.X: 134542334 - 134583623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013482.2 NP_038510.2 14 - 15 1504 74
GenBank mRNA BC053392.1 - 14 - 15 1440 74
L08967.1 - 14 - 15 1480 74
L29788.1 - 14 - 15 1476 74

View Full Product Details