Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr4
Entrez Gene ID
57773
Gene Name
WD repeat domain 4
Gene Aliases
AI415180, AI448349, D530049K22Rik
Chromosome Location
Chr.17: 31494322 - 31519992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021322.2 NP_067297.2 1 - 2 320 67
GenBank mRNA AK035125.1 - 1 - 2 101 67
AK144821.1 - 1 - 2 125 67
AK152271.1 - 1 - 2 112 67
AK153161.1 - 1 - 2 112 67
AK164160.1 - 1 - 2 85 67
BC039272.1 - 1 - 2 120 67
CD578615.1 - 3 - 4 408 67

View Full Product Details