The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044384.1 | NP_001037849.1 | 1 - 2 | 157 | 87 |
NM_001294280.2 | NP_001281209.1 | 1 - 2 | 157 | 87 | |
GenBank mRNA | BC008107.1 | - | 1 - 2 | 59 | 87 |
BC034260.1 | - | 1 - 2 | 59 | 87 | |
CJ055052.1 | - | 1 - 2 | 69 | 87 |