Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gadl1
Entrez Gene ID
73748
Gene Name
glutamate decarboxylase-like 1
Gene Aliases
1110027M19Rik, ADC, CSADC
Chromosome Location
Chr.9: 115884644 - 116076176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028638.1 NP_082914.1 8 - 9 811 121
GenBank mRNA BC052327.1 - 8 - 9 811 121

View Full Product Details