Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2d5
Entrez Gene ID
230863
Gene Name
SH2 domain containing 5
Gene Aliases
BC036961
Chromosome Location
Chr.4: 138250411 - 138260968 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099631.1 NP_001093101.1 4 - 5 381 113
GenBank mRNA AK032598.1 - 4 - 5 315 113
BC036961.1 - 5 - 6 415 113
DQ832275.1 - 4 - 5 381 113

View Full Product Details