Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrto4
Entrez Gene ID
69902
Gene Name
MRT4, mRNA turnover 4, homolog (S. cerevisiae)
Gene Aliases
2610012O22Rik, Mg684, Mrt4
Chromosome Location
Chr.4: 139347440 - 139352576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023536.3 NP_076025.1 7 - 8 854 121
GenBank mRNA BC005734.1 - 7 - 8 576 121

View Full Product Details