Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hk3
Entrez Gene ID
212032
Gene Name
hexokinase 3
Gene Aliases
HK III, HK-III
Chromosome Location
Chr.13: 55005985 - 55021472 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033245.4 NP_001028417.1 18 - 19 2890 70
NM_001206390.1 NP_001193319.1 18 - 19 2700 70
NM_001206391.1 NP_001193320.1 18 - 19 2565 70
NM_001206392.1 NP_001193321.1 17 - 18 2770 70
GenBank mRNA AK140444.1 - 18 - 19 2754 70
AK149651.1 - 18 - 19 2740 70
AK149992.1 - 18 - 19 2702 70
AK154603.1 - 18 - 19 2862 70
AK159857.1 - 17 - 18 2616 70
AK159906.1 - 18 - 19 2800 70
AK160018.1 - 18 - 19 2800 70
AK160063.1 - 18 - 19 2800 70
AK162637.1 - 18 - 19 2833 70
AK163658.1 - 18 - 19 2754 70
AK170813.1 - 17 - 18 2620 70
AK171214.1 - 18 - 19 2828 70
AK171248.1 - 18 - 19 2740 70
AK171580.1 - 18 - 19 2840 70
BC117860.1 - 18 - 19 2785 70
BC117861.1 - 18 - 19 2650 70
BI133441.1 - 1 - 2 70

View Full Product Details