Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tekt5
Entrez Gene ID
70426
Gene Name
tektin 5
Gene Aliases
3300001K11Rik
Chromosome Location
Chr.16: 10357952 - 10395465 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099275.1 NP_001092745.1 4 - 5 894 59
NM_001291001.1 NP_001277930.1 4 - 5 911 59
GenBank mRNA BC115969.1 - 4 - 5 894 59

View Full Product Details