The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001101588.1 | NP_001095058.1 | 9 - 10 | 1254 | 74 |
GenBank mRNA | BC158089.1 | - | 9 - 10 | 1331 | 74 |