Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upk1b
Entrez Gene ID
22268
Gene Name
uroplakin 1B
Gene Aliases
AI413235, Tspan20, UPIb, Upk1
Chromosome Location
Chr.16: 38773184 - 38800345 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178924.4 NP_849255.2 3 - 4 348 99
GenBank mRNA AF274865.1 - 2 - 3 283 99
AK053004.1 - 3 - 4 349 99
AK083101.1 - 3 - 4 348 99
AK086926.1 - 3 - 4 347 99
BC046604.1 - 3 - 4 326 99

View Full Product Details