The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178924.4 | NP_849255.2 | 7 - 8 | 805 | 62 |
GenBank mRNA | AF274865.1 | - | 6 - 7 | 740 | 62 |
AK053004.1 | - | 7 - 8 | 806 | 62 | |
AK083101.1 | - | 7 - 8 | 805 | 62 | |
AK086926.1 | - | 7 - 8 | 804 | 62 | |
BC046604.1 | - | 7 - 8 | 783 | 62 |