Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt10
Entrez Gene ID
171212
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
AU018154, C330012K04Rik, GalNAc-T10, GalNAc-T9, Galnt9, ppGaNTase
Chromosome Location
Chr.11: 57622407 - 57787501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134189.2 NP_598950.2 2 - 3 376 71
GenBank mRNA AK131155.1 - 2 - 3 354 71
AK143714.1 - 2 - 3 374 71
AK159362.1 - 2 - 3 374 71
AK162424.1 - 2 - 3 376 71
AK172609.1 - 2 - 3 356 71
BB626609.1 - 2 - 3 376 71
BC060617.1 - 2 - 3 359 71

View Full Product Details