Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Galnt10
Entrez Gene ID
171212
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 10
Gene Aliases
AU018154, C330012K04Rik, GalNAc-T10, GalNAc-T9, Galnt9, ppGaNTase
Chromosome Location
Chr.11: 57622407 - 57787501 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134189.2 NP_598950.2 3 - 4 513 112
GenBank mRNA AK033515.1 - 2 - 3 171 112
AK131155.1 - 3 - 4 491 112
AK159088.1 - 3 - 4 512 112
AK159362.1 - 3 - 4 511 112
AK162424.1 - 3 - 4 513 112
BB626609.1 - 3 - 4 513 112
BC060617.1 - 3 - 4 496 112

View Full Product Details