The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011672.4 | NP_035802.3 | 1 - 2 | 120 | 63 |
NR_028403.1 | 1 - 2 | 120 | 63 | ||
GenBank mRNA | AK010461.1 | - | 1 - 2 | 89 | 63 |
AK088823.1 | - | 1 - 2 | 89 | 63 | |
AK147138.1 | - | 1 - 2 | 103 | 63 | |
AK169062.1 | - | 1 - 2 | 102 | 63 | |
BC006630.1 | - | 1 - 2 | 77 | 63 | |
CJ053815.1 | - | 1 - 2 | 115 | 63 | |
CJ085208.1 | - | 1 - 2 | 120 | 63 | |
U64445.1 | - | 1 - 2 | 105 | 63 |