The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178720.4 | NP_848835.1 | 11 - 12 | 1229 | 59 |
GenBank mRNA | AK034628.1 | - | 11 - 12 | 1230 | 59 |
AK047981.1 | - | 10 - 11 | 1139 | 59 | |
AK052599.1 | - | 12 - 13 | 1339 | 59 | |
BC116255.1 | - | 11 - 12 | 1198 | 59 |