Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zbtb20
- Entrez Gene ID
-
56490
- Gene Name
- zinc finger and BTB domain containing 20
- Gene Aliases
-
1300017A20Rik, 7330412A13Rik, A930017C21Rik, D16Wsu73e, DPZF, HOF, ODA-8S, Oda8, Zfp288
- Chromosome Location
-
Chr.16: 42875853 - 43621944 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK078664.1 |
- |
- |
96 |
69 |
BI659024.1 |
- |
3 - 4 |
176 |
69 |
View Full Product Details