The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012765.4 | NP_001012783.3 | 15 - 16 | 3367 | 62 |
GenBank mRNA | AK147590.1 | - | - | 3470 | 62 |
AK147649.1 | - | - | 4127 | 62 | |
AK160942.1 | - | 15 - 16 | 3367 | 62 | |
BC035550.1 | - | - | 1505 | 62 | |
BC090846.1 | - | 15 - 16 | 3102 | 62 |