Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adcy5
Entrez Gene ID
224129
Gene Name
adenylate cyclase 5
Gene Aliases
AW121902, Ac5
Chromosome Location
Chr.16: 35155636 - 35304549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012765.4 NP_001012783.3 1 - 2 1601 85
GenBank mRNA AK147590.1 - - 1702 85
AK147649.1 - - 2361 85
AK160942.1 - 1 - 2 1601 85
BC090846.1 - 1 - 2 1336 85

View Full Product Details