Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gbe1
Entrez Gene ID
74185
Gene Name
glucan (1,4-alpha-), branching enzyme 1
Gene Aliases
2310045H19Rik, 2810426P10Rik, D16Ertd536e
Chromosome Location
Chr.16: 70313949 - 70569720 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028803.4 NP_083079.1 13 - 14 2008 148
GenBank mRNA AK009815.1 - 13 - 14 1824 148
AK050365.1 - 13 - 14 1895 148
AK050423.1 - 13 - 14 1894 148
AK148186.1 - 7 - 8 936 148
BC017541.1 - 13 - 14 1869 148

View Full Product Details