Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fetub
Entrez Gene ID
59083
Gene Name
fetuin beta
Gene Aliases
2310011O17Rik, AI255764, D17980, Gugu
Chromosome Location
Chr.16: 22918382 - 22939768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083904.1 NP_001077373.1 2 - 3 387 115
NM_001083905.1 NP_001077374.1 3 - 4 557 115
NM_021564.2 NP_067539.1 3 - 4 622 115
GenBank mRNA AJ242927.1 - 3 - 4 543 115
AK009310.1 - 3 - 4 611 115
AK009554.1 - 3 - 4 556 115
AY138459.1 - 3 - 4 607 115
AY138460.1 - 2 - 3 157 115
BC018341.1 - 3 - 4 487 115

View Full Product Details