Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap4
Entrez Gene ID
83383
Gene Name
transcription factor AP4
Gene Aliases
AI642933, AP-4, D930048N17Rik, Tcfap4, bHLHc41
Chromosome Location
Chr.16: 4544661 - 4560772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031182.2 NP_112459.1 5 - 6 867 165
GenBank mRNA AF161262.1 - 5 - 6 867 165
AK053091.1 - 5 - 6 659 165
BC047270.1 - 5 - 6 774 165
BC054777.1 - 5 - 6 758 165

View Full Product Details