Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jam2
Entrez Gene ID
67374
Gene Name
junction adhesion molecule 2
Gene Aliases
1110002N23Rik, 2410030G21Rik, 2410167M24Rik, JAM-2, JAM-B, Jcam2, VE-JAM
Chromosome Location
Chr.16: 84774123 - 84826380 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023844.5 NP_076333.3 9 - 10 1219 95
GenBank mRNA AF255911.1 - 9 - 10 942 95
AK010616.1 - 9 - 10 1061 95
AK013914.1 - 9 - 10 1060 95
AK028757.1 - 9 - 10 910 95
AK078128.1 - 9 - 10 945 95
AK153636.1 - 9 - 10 910 95
BC028778.1 - 9 - 10 1219 95

View Full Product Details