Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 26 - 27 3658 81
NM_013607.2 NP_038635.2 26 - 27 3658 81
GenBank mRNA AK137226.1 - 27 - 28 3635 81
AK173045.1 - 26 - 27 3622 81
BC026142.1 - 26 - 27 3643 81
D85923.1 - 26 - 27 3613 81
D85924.1 - 26 - 27 3613 81

View Full Product Details