Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pigx
Entrez Gene ID
72084
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class X
Gene Aliases
2010319C14Rik, PIG-X
Chromosome Location
Chr.16: 32084416 - 32099727 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111025.1 NP_001104495.1 2 - 3 315 106
NM_024464.3 NP_077784.2 2 - 3 321 106
GenBank mRNA AK008583.2 - 2 - 3 323 106
BX525135.1 - 2 - 3 288 106
BY708475.1 - 2 - 3 322 106

View Full Product Details