The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113208.1 | NP_001106679.1 | 14 - 15 | 2112 | 125 |
NM_010954.4 | NP_035084.1 | 14 - 15 | 2112 | 125 | |
GenBank mRNA | AF001286.1 | - | 14 - 15 | 2038 | 125 |
AF001287.1 | - | 14 - 15 | 2038 | 125 | |
AF016619.1 | - | 14 - 15 | 1994 | 125 | |
BC119027.1 | - | 14 - 15 | 1970 | 125 | |
BC119029.1 | - | 14 - 15 | 1970 | 125 |