Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parp14
Entrez Gene ID
547253
Gene Name
poly (ADP-ribose) polymerase family, member 14
Gene Aliases
1600029O10Rik, ARTD8, BC021340, CoaSt6, mKIAA1268
Chromosome Location
Chr.16: 35832859 - 35871514 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039530.3 NP_001034619.2 14 - 15 4890 61
GenBank mRNA AK220263.1 - 9 - 10 2482 61
BC021340.1 - 2 - 3 741 61
BC047386.1 - 9 - 10 1990 61
BC138874.1 - 14 - 15 5005 61
BC145452.1 - 13 - 14 5092 61
DQ372930.1 - 14 - 15 5054 61

View Full Product Details