Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zbtb11
- Entrez Gene ID
-
271377
- Gene Name
- zinc finger and BTB domain containing 11
- Gene Aliases
-
9230110G02Rik, ZNF-U69274
- Chromosome Location
-
Chr.16: 55972697 - 56008913 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_173026.2 |
NP_766614.2 |
3 - 4 |
1032 |
115 |
View Full Product Details