Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
27886
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
AI462402, D16H22S1269E, Dgcr1, Dgsi, ES2, Es2el
Chromosome Location
Chr.16: 17900709 - 17911364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022408.2 NP_071853.2 3 - 4 456 84
GenBank mRNA AF037256.1 - 3 - 4 455 84
AK142684.1 - 3 - 4 421 84

View Full Product Details