Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
27886
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
AI462402, D16H22S1269E, Dgcr1, Dgsi, ES2, Es2el
Chromosome Location
Chr.16: 17900709 - 17911364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081633.1 NP_001075102.1 7 - 8 981 69
NM_022408.2 NP_071853.2 7 - 8 984 69
GenBank mRNA AF037256.1 - 7 - 8 983 69
AK088520.1 - 7 - 8 946 69
AK142684.1 - 7 - 8 949 69
AK148404.1 - 7 - 8 952 69
BC013711.1 - 7 - 8 960 69

View Full Product Details