The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081633.1 | NP_001075102.1 | 9 - 10 | 1204 | 58 |
NM_022408.2 | NP_071853.2 | 9 - 10 | 1207 | 58 | |
GenBank mRNA | AF037256.1 | - | 9 - 10 | 1206 | 58 |
AK088520.1 | - | 9 - 10 | 1169 | 58 | |
AK142684.1 | - | 9 - 10 | 1172 | 58 | |
AK148404.1 | - | 9 - 10 | 1175 | 58 | |
BC013711.1 | - | 9 - 10 | 1183 | 58 |