Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gsc2
Entrez Gene ID
195333
Gene Name
goosecoid homebox 2
Gene Aliases
4930568H22Rik, GSC-2, Gscl
Chromosome Location
Chr.16: 17913554 - 17915059 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029469.1 NP_083745.1 1 - 2 306 94
GenBank mRNA BC125356.1 - 1 - 2 306 94
BC132637.1 - 1 - 2 306 94

View Full Product Details