The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019494.1 | NP_062367.1 | 3 - 4 | 318 | 88 |
NR_038116.1 | 3 - 4 | 336 | 88 | ||
GenBank mRNA | AF136449.1 | - | 3 - 4 | 318 | 88 |
AF178672.1 | - | 3 - 4 | 318 | 88 | |
AF179872.1 | - | 3 - 4 | 318 | 88 | |
AK020750.1 | - | 3 - 4 | 337 | 88 | |
AK040051.1 | - | 3 - 4 | 278 | 88 | |
AK050012.1 | - | - | 2525 | 88 | |
BC025903.1 | - | 3 - 4 | 306 | 88 |